Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN28 1632, >98%, Tocris Bioscience™
RNA binding protein Lin28 inhibitor; promotes mESC differentiation; also bromodomain inhibitor
$251.72 - $1075.32
Specifications
Chemical Name or Material | N-Methyl-N-[3-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide |
---|---|
CAS | 108825-65-6 |
Purity Grade Notes | >98% |
Linear Formula | C15H15N5O |
Solubility Information | Soluble to 100mM in DMSO and to 100mM in ethanol |
Description
RNA binding protein Lin28 inhibitor (IC50 = 8 μM). Blocks binding of Lin28 to let-7 pre-micro RNA. Inhibits stemness and induces differentiation of murine ESCs. Inhibits growth of tumor cell lines and tumor-sphere formation in vitro. Also bromodomain inhibitor.Specifications
N-Methyl-N-[3-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide | |
>98% | |
Soluble to 100mM in DMSO and to 100mM in ethanol |
108825-65-6 | |
C15H15N5O | |
281.31 |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title