Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LIN28 1632, >98%, Tocris Bioscience™
RNA binding protein Lin28 inhibitor; promotes mESC differentiation; also bromodomain inhibitor
Supplier: R&D Systems 6068/50
This item is not returnable.
View return policy
Description
Lin28 1632 is an RNA binding protein Lin28 inhibitor (IC50 = 8 μM). Lin28 1632 blocks binding of Lin28 to let-7 pre-micro RNA. Lin28 1632 inhibits stemness and induces differentiation of murine ESCs. Lin28 1632 increases let-7 miRNA and suppresses PD-L1 expression in multiple human cancer cell lines. Lin28 1632 inhibits growth of tumor cell lines and tumor-sphere formation in vitro. Lin28 1632 also inhibits growth of tumor xenografts in mice by enhancing T cell-mediated antitumor immunity . Lin28 1632 is also bromodomain inhibitor. Lin28 1632 inhibits tumor growth and metastasis in vivo in immunodeficient orthotopic mouse models of human breast cancer cell MDA-MB-231.Specifications
N-Methyl-N-[3-(3-methyl-1,2,4-triazolo[4,3-b]pyridazin-6-yl)phenyl]acetamide | |
>98% | |
50 mg | |
281.31 |
108825-65-6 | |
C15H15N5O | |
Soluble to 100mM in DMSO and to 100mM in ethanol |
Safety and Handling
Recommended Storage : Store at 4°C
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction