Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reagents
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
Specifications
Chemical Name or Material | Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard |
---|---|
CAS | 7732-18-5, 877-24-7 |
Melting Point | 0°C |
Boiling Point | 100°C |
Color | Pink |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
01-337-438
|
Reagents Holdings Llc
C2155500100S |
100 mL | Poly Bottle |
Each for $8.06
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-437
|
Reagents Holdings Llc
C215550010F |
10 L | Cubitainer |
Each for $128.24
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-436
|
Reagents Holdings Llc
C21555001A |
1 L | Poly Bottle |
Each for $39.58
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-435
|
Reagents Holdings Llc
C2155500200E |
200 L | Poly Drum |
Each for $570.32
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-434
|
Reagents Holdings Llc
C215550020F |
20 L | Cubitainer |
Each for $161.82
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-433
|
Reagents Holdings Llc
C21555004A |
4 L | Poly Bottle |
Each for $81.80
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-432
|
Reagents Holdings Llc
C21555004F |
4 L | Cubitainer |
Each for $84.72
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
01-337-431
|
Reagents Holdings Llc
C2155500500A |
500 mL | Poly Bottle |
Each for $25.18
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Specifications
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard | |
0°C | |
Pink | |
pH Meter | |
Liquid | |
1.0039 |
7732-18-5, 877-24-7 | |
100°C | |
Calibration | |
4 | |
Buffer | |
Non-sterile |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title