Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reagents

Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Supplier:  Reagents Holdings Llc C21555004A

 View more versions of this product

Catalog No. 01-337-433


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Specifications

Specifications

Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard
0°C
Pink
pH Meter
Liquid
Buffer
1.0039
7732-18-5, 877-24-7
100°C
Calibration
4
4 L
Poly Bottle
Non-sterile
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit