Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reagents
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
Supplier: Reagents Holdings Llc C215550010F
Specifications
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard | |
0°C | |
Pink | |
pH Meter | |
Liquid | |
Buffer | |
1.0039 |
7732-18-5, 877-24-7 | |
100°C | |
Calibration | |
4 | |
10 L | |
Cubitainer | |
Non-sterile |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction