Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reagents
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard, Reference Buffer, use for pH meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
Supplier: Reagents Holdings Llc C2155500100S
Specifications
Buffer pH 4.00 ± 0.01 at 25°C, Color-Coded Red Reference Standard | |
0°C | |
Pink | |
pH Meter | |
Liquid | |
Buffer | |
1.0039 |
7732-18-5, 877-24-7 | |
100°C | |
Calibration | |
4 | |
100 mL | |
Poly Bottle | |
Non-sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction