Learn More
Dentec Safety™ Comfort-Air™NxMD Filter Cover For N95 Filter Pads
SDP
Dentec Comfort-Air™NxMD filter cover for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
$22.80 - $555.50
Specifications
Disposable Reusable
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
19-222-568
SDP
View Documents
Dentec Safety™
14668W6
6 Pk.
Pack of 6 for $22.80
Add to Cart
19-222-569
SDP
View Documents
Dentec Safety™
14668WB350
350/Cs.
Case of 350 for $555.50
Add to Cart
Description
Description
  • Filter Cover
  • Allows user to conduct a positive and negative seal check
  • Protects N95 filters from damage
  • Extends filter life
  • Prevents the user from touching the filter
  • Rugged and durable
  • Can be easily disinfected
Specifications
Specifications
Reusable