Learn More
Dentec Safety™ Comfort-Air™NxMD Filter Cover For N95 Filter Pads
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Dentec Comfort-Air™NxMD filter cover for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
Supplier: Dentec Safety™ 14668WB350
Description
- Filter Cover
- Allows user to conduct a positive and negative seal check
- Protects N95 filters from damage
- Extends filter life
- Prevents the user from touching the filter
- Rugged and durable
- Can be easily disinfected
Specifications
Reusable | |
350/Cs. |