Learn More
Dentec Safety™ Comfort-Air™NxMD Filter Cover For N95 Filter Pads
SDP
Dentec Comfort-Air™NxMD filter cover for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
Supplier:  Dentec Safety™ 14668W6
 View more versions of this product
Catalog No. 19-222-568
Add to Cart
Description
Description
  • Filter Cover
  • Allows user to conduct a positive and negative seal check
  • Protects N95 filters from damage
  • Extends filter life
  • Prevents the user from touching the filter
  • Rugged and durable
  • Can be easily disinfected
Specifications
Specifications
Reusable
6 Pk.
Product Suggestions
Product Suggestions