Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Terpinolene, Natural Isolate, Ricca Chemical
Terpinolene, Natural Isolate is a natural isolate food additive.
$16.25 - $151.00
Specifications
Chemical Name or Material | Terpinolene |
---|---|
CAS | 586-62-9 |
Packaging | Amber Glass |
Color | Colorless to Light Yellow |
Certificate of Analysis | Yes |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-184-934
|
Ricca Chemical Company
RCCT900220C6 |
20 g |
Each for $16.25
|
|
|||||
01-184-917
|
Ricca Chemical Company
RCCT9002100C6 |
100 g |
Each for $33.25
|
|
|||||
01-184-937
|
Ricca Chemical Company
RCCT9002750C6 |
750 g |
Each for $151.00
|
|
|||||
Description
- All Natural
- Food Grade
- FSSC 22000 and ISO 9001 Accreditated Facility
- Additional Food Safety Testing Included (Pesticide, Residual Solvents, Heavy Metals)
Specifications
Terpinolene | |
Amber Glass | |
Yes | |
90% to 100% | |
136.24 | |
Piney, mild, sweet, citrus |
586-62-9 | |
Colorless to Light Yellow | |
Food Additive Standard | |
C10H16 | |
Food Grade | |
Liquid |
Safety and Handling
ShelfLife : 24 Months
Recommended Storage : 15°C to 30°C (59°F to 86°F)
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title