Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Terpinolene, Natural Isolate, Ricca Chemical
Terpinolene, Natural Isolate is a natural isolate food additive.
Supplier: Ricca Chemical Company RCCT9002100C6
Description
- All Natural
- Food Grade
- FSSC 22000 and ISO 9001 Accreditated Facility
- Additional Food Safety Testing Included (Pesticide, Residual Solvents, Heavy Metals)
Specifications
Terpinolene | |
586-62-9 | |
Colorless to Light Yellow | |
Food Additive Standard | |
C10H16 | |
Food Grade | |
Liquid |
Amber Glass | |
100 g | |
Yes | |
90% to 100% | |
136.24 | |
Piney, mild, sweet, citrus |
Safety and Handling
ShelfLife : 24 Months
Recommended Storage : 15°C to 30°C (59°F to 86°F)
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction