Learn More
3M™ Standard Cable Ties (50lb, 220N) UL Listed TYPE 21S
3M™ Cable Ties are used to secure wire bundles and harness components quickly. The flexible design allows the user to slip ties easily under and around cables and harnesses.
$123.50 - $1034.00
Specifications
Material | Nylon 6/6 |
---|---|
Tensile Strength (English) | 50 lb. |
Width (English) | 0.18 in. |
Width (Metric) | 4.6 mm |
Catalog Number | Mfr. No. | Color | Length (English) | Product Type | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Length (English) | Product Type | Quantity | Price | Quantity & Availability | ||
19-071-011
|
3M™
7000031853 |
Black | 7.6 in. | Standard Cable Tie | 1000/Cs. |
Case of 1000 for $143.20
|
|
||
19-071-051
|
3M™
7000031854 |
Black | 7.6 in. | Standard Cable Tie | 10,000/Cs. |
Case of 10000 for $1,034.00
|
|
||
19-071-036
|
3M™
7000058850 |
Black | 11.1 in. | Standard Cable Tie | 1000/Cs. |
Case of 1000 for $201.50
|
|
||
19-071-014
|
3M™
7000031857 |
Black | 14.6 in. | Standard Cable Tie | 500/Cs. |
Case of 500 for $130.70
|
|
||
19-071-042
|
3M™
7000031858 |
Black | 14.6 in. | Standard Cable Tie | 2500/Cs. |
Case of 2500 for $581.00
|
|
||
19-071-012
|
3M™
7000133461 |
Black | 14.6 in. | Standard Cable Tie | 5000/Cs. |
Case of 5000 for $815.50
|
|
||
19-071-054
|
3M™
7000031855 |
Natural | 7.6 in. | Miniature Cable Tie | 1000/Cs. |
Case of 1000 for $128.80
|
|
||
19-071-041
|
3M™
7000133460 |
Natural | 7.6 in. | Standard Cable Tie | 10,000/Cs. |
Case of 10000 for $917.00
|
|
||
19-071-038
|
3M™
7000031856 |
Natural | 11.1 in. | Miniature Cable Tie | 1000/Cs. |
Case of 1000 for $178.40
|
|
||
19-071-013
|
3M™
7000133462 |
Natural | 11.1 in. | Standard Cable Tie | 5000/Cs. |
Case of 5000 for $742.50
|
|
||
19-071-016
|
3M™
7000031859 |
Natural | 14.6 in. | Standard Cable Tie | 500/Cs. |
Case of 500 for $123.50
|
|
||
19-071-018
|
3M™
7000133463 |
Natural | 14.6 in. | Standard Cable Tie | 2500/Cs. |
Case of 2500 for $521.00
|
|
||
19-071-029
|
3M™
7010400149 |
Red | 7.6 in. | Standard Cable Tie | 1000/Cs. |
Case of 1000 for $143.20
|
|
||
19-071-026
|
3M™
7010349835 |
Red | 11.1 in. | Standard Cable Tie | 1000/Cs. |
Case of 1000 for $201.50
|
|
||
Description
- Self-locking heads lock in any position once threaded
- Harness components and tie down wire bundles quickly
- Designed to function in a wide temperature range between -40 to 185°F (-40 to 85°C)
- Used to bind wires, cables and cords together securely
- 3M Catalog# CT11BK50-C, CT11BK50-D, CT11NT50-C, CT11NT50-D, CT11RD50-C, CT15BK50-C, CT15BK50-D, CT15NT50-C, CT15NT50-D, CT8BK50-C, CT8BK50-M, CT8NT50-C, CT8NT50-M, CT8RD50-C
Specifications
Nylon 6/6 | |
50 lb. | |
0.18 in. | |
4.6 mm |