Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse SCF/c-kit Ligand Protein
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Recombinant Protein
$248.86 - $3192.05
Specifications
For Use With (Application) | Bioactivity, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot |
---|---|
Formulation | Lyophilized from a 0.2 micron filtered solution, 20 mM sodium phosphate, pH 6.5 |
Gene ID (Entrez) | 4254 |
Storage Requirements | Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. |
Endotoxin Concentration | Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB171767
![]() |
Novus Biologicals™
NBP318185500UG |
500 μg |
Each for $1,907.95
|
|
|||||
NB171765
![]() |
Novus Biologicals™
NBP3181851MG |
1 mg |
Each for $3,192.05
|
|
|||||
NB171764
![]() |
Novus Biologicals™
NBP31818510UG |
10 μg |
Each for $248.86
|
|
|||||
NB171763
![]() |
Novus Biologicals™
NBP318185100UG |
100 μg |
Each for $589.77
|
|
|||||
Description
TF-1 cell proliferation, >5.0 x 10^4 units/mg (ED50 acceptance criteria are <20 ng/mL).Specifications
Bioactivity, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot | |
4254 | |
Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). | |
KITLG | |
Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
SCF/c-kit Ligand |
Lyophilized from a 0.2 micron filtered solution, 20 mM sodium phosphate, pH 6.5 | |
Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. | |
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
Sterile water at 0.1 mg/mL | |
>95%, by SDS-PAGE |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title