Learn More
Novus Biologicals™ Recombinant Mouse SCF/c-kit Ligand Protein
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Recombinant Protein
Supplier: Novus Biologicals™ NBP318185100UG
This item is not returnable.
View return policy
Description
TF-1 cell proliferation, >5.0 x 10^4 units/mg (ED50 acceptance criteria are <20 ng/mL).Specifications
Bioactivity, Immunohistochemistry, Immunohistochemistry-Paraffin, SDS-PAGE, Western Blot | |
4254 | |
Store at -20 to -70C as supplied. After reconstitution, store at 2 to 8C for 1 month and at -20 to -70C for long term storage. Avoid repeated freeze-thaw cycles. | |
DKFZp686F2250, familial progressive hyperpigmentation 2, FPH2, KIT ligand, Kitl, KL-1, Mast cell growth factor, MGFSHEP7, SCFStem cell factor, SFc-Kit ligand, steel factor | |
Sterile water at 0.1 mg/mL | |
>95%, by SDS-PAGE |
Lyophilized from a 0.2 micron filtered solution, 20 mM sodium phosphate, pH 6.5 | |
100 μg | |
Acceptance criteria for endotoxin levels are <1 EUs/μg protein (determined by Kinetic LAL Method). | |
KITLG | |
Biologically Active Proteins, Cancer, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
SCF/c-kit Ligand |