Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NS 9283
Positive allosteric modulator of α4β2 nAChRs
$291.16 - $1212.20
Specifications
Purity | >99% |
---|---|
Product Type | NS 9283 |
Molecular Weight (g/mol) | 248.24 |
Inhibitors | NS 9283 |
Description
Positive allosteric modulator of α4β2 receptor; increases the potency of Ach-evoked currents ∽60 fold without effecting the maximum efficacy (HEK293-hα4β2 cells). Reduces the rate of recovery from desensitization and slows the rate of deactivation. Displays a synergistic effect on the Emax when given with NS206 at 3α:2β receptors.Specifications
>99% | |
NS 9283 | |
248.24 | |
NS 9283 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title