Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lonza™ BioWhittaker™ L-15 (Leibovitz) Medium

Supplier:  Lonza™ 12700F

 View more versions of this product

Catalog No. BW12-700F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Explore available promotions
This item is not returnable. View return policy

Description

Description

  • Manufactured using formulations recommended by the Tissue Culture Association and Water for Injection (WFI) quality water
  • Sterile-filtered through two consecutive 0.2μm pharmaceutical-grade filters

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C
Sterile
No Glutamine
500 mL
Leibovitz's L-15
Sterile-filtered through two consecutive 0.2 μm filters
7.3 to 7.75
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.