Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LM11A 31 dihydrochloride
Nonpeptide p75NTR ligand; neuroprotective
$169.36 - $700.64
Specifications
Purity | 95% |
---|---|
Molecular Formula | (2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride |
CAS | 1243259-19-9 |
Content And Storage | Desiccate at RT |
Product Type | LM11A 31 dihydrochloride |
Description
Nonpeptide p75NTR ligand; blocks p75-mediated cell death and also increases proliferation and survival of hippocampal neural progenitors. Exhibits no effect on nerve growth factor (NGF) binding to TrkA. Prevents and reverses atrophy of cholinergic neurites, as well as reversing Alzheimer's Disease (AD) pathologies in mid- to late-stage AD mice models. Shown to promote functional recovery in a mouse model of spinal cord injury. Orally available and brain penetrant.Specifications
95% | |
1243259-19-9 | |
LM11A 31 dihydrochloride | |
LM11A 31 dihydrochloride |
(2 S,3 S)-2-Amino-3-methyl-N-[2-(4-morpholinyl)ethyl]pentanamide dihydrochloride | |
Desiccate at RT | |
M.W. (Observed): 316.27 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title