Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Fritted Hirsch Filter Funnels
Directs filtrate through a reduced area frit to concentrate crystal residue for rewashing
Supplier: DWK Life Sciences 9552507523
Description
- Conical shape
- Available in two porosities: fine and coarse
Specifications
8 mm | |
145 mm | |
Directs filtrate through a reduced area frit to concentrate crystal residue for rewashing | |
75 mm | |
40 to 60 μm |
Borosilicate Glass | |
USP Type I, ASTM E438, Class A | |
30 mm | |
Coarse | |
Hirsch Funnel |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction