Learn More
Dentec Safety™ Comfort-Air™NxMD Filter Holder For N95 Filter Pads
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Dentec Comfort-Air™NxMD NxMD filter holder for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
$30.45 - $533.00
Specifications
Disposable | Reusable |
---|---|
Product Type | Filter Holder |
With Exhalation Valve | No |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-222-566
![]() |
Dentec Safety™
14880W |
6 Pk. |
Pack of 6 for $30.45
|
|
|||||
19-222-567
![]() |
Dentec Safety™
14880WB350 |
350/Cs. |
Case of 350 for $533.00
|
|
|||||
Description
- Ultra thin design will fit under any grinding shield or welding helmet
- Will not obstruct field of vision
- Holder has spacers that create a space between the filter material and the holder
- This allows the user to access the entire filter material increasing filter life while reducing breathing resistance and increasing productivity
Specifications
Reusable | |
Filter Holder | |
No |