Learn More
Dentec Safety™ Comfort-Air™NxMD Filter Holder For N95 Filter Pads
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Dentec Comfort-Air™NxMD NxMD filter holder for N95 filter pads. Suitable for COVID virus in accordance with CDC guidelines and non-oil based aerosols. Can be mounted directly on all Comfort-Air™ series half and full face masks.
Supplier: Dentec Safety™ 14880WB350
Description
- Ultra thin design will fit under any grinding shield or welding helmet
- Will not obstruct field of vision
- Holder has spacers that create a space between the filter material and the holder
- This allows the user to access the entire filter material increasing filter life while reducing breathing resistance and increasing productivity
Specifications
Reusable | |
No | |
350/Cs. |