Learn More
TARGETMOL CHEMICALS INC ZEARALENONE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T5625
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Zearalenone (F2 toxin) is a mycotoxin produced mainly by fungi belonging to the genus Fusarium in foods and feeds. Possess oestrogenic activity in cattle pigs and sheep with low acute toxicity. Causes precocious development of mammae and other estrogenic effects in young gilts. purity: 99%
Catalog No. 50-257-9745
Explore available promotions