Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HORIBA Yumizen C1200 U/CSF Micros Alb Control
Yumizen C1200 U/CSF Micros Alb Control used to monitor micro albumin in urine
Specifications
Certifications/Compliance | CE Marked |
---|---|
Content And Storage | 2°C to 8°C |
Concentration | Normal concentration |
Detectable Analytes | Microalbumin |
For Use With (Equipment) | Used on Yumizen C1200 Clinical Chemistry Analyer |
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Level | Price | Quantity & Availability | |||||
23-601-594
|
HORIBA
1300023948 |
Normal Level |
Each for $126.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-601-595
|
HORIBA
1300023949 |
Pathological Level |
Each for $134.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Used to monitor micro albumin in urine
Specifications
CE Marked | |
Normal concentration | |
Used on Yumizen C1200 Clinical Chemistry Analyer | |
Assay Control | |
Yes | |
Human Blood Material (Plasma) | |
Liquid |
2°C to 8°C | |
Microalbumin | |
Plasma | |
Control | |
3 Pk. 1 mL Vials | |
Stable for 30 days (at 2°C to 8°C) |
Safety and Handling
ShelfLife : 10 months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title