Learn More
High Purity Standards YTTRIUM AT 1000PPM IN 2% HNO3
Supplier: High Purity Standards 100067150
Y 1000ug/mL in 2% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.