Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
YSI™ TruLab™ pH/ISE 1320P Dual Channel Laboratory ISE Benchtop Meter
Easy-to-use meter with integrated thermal printer for dual measurement of pH, mV (ORP) or ISE and temperature
Supplier: YSI™ 1GA340PY
Includes: Meter with integrated printer, universal power supply, electrode stand, manual, PC software and USB cable
Description
- Provides automatic documentation for GLP compliance
- AutoRead function improves repeatability
- Easy-to-read backlit display with text menu for easy operation for pH, mV (ORP), ISE and temperature
- Fast data transfer via USB in .csv or ASCII format or print with integrated thermal printer
- Easy calibration with adjustable calibration timer which indicates when to perform the next calibration routine
- Automatic or manual temperature compensation
- 1, 2, 3, 4 or 5 point calibration for pH
- 1, 2, 3, 4, 5, 6 or 7 point calibration for ISE, also non- linear
- BNC connection for pH, mV and ISEs; banana connection for temperature probes (sold separately)
- Continuous Monitoring Control (CMC) function for monitoring the ideal measuring range for pH measurements based on the last calibration
- Wide selection of electrodes: TruLab analog pH/mV/ISE/temp probes
- Four alkaline AA batteries provide approx.150 hours with illumination; universal power supply; also compatible with four NiMH AA rechargeables
Parameters Measured:
- pH, mV (ORP/Redox)
- Temperature
- ISEs: Ammonia, Ammonium, Bromide, Cadmium, Calcium, Chloride, Copper, Cyanide, Fluoride, Iodide, Lead, Nitrate, Potassium, Silver/Sulfide, Sodium
Specifications:
- Buffer Recognition: Yes
- Sampling: Yes
- User Calibratable: Yes
- Logging Capabilities: Yes
- pH Range: -2.0 to 20.0; -2.00 to 20.00; -2.000 to 19.999
- pH Resolutions: 0.1; 0.01; 0.001
- pH Accuracy (±1 digit): ±0.1; ±0.01; ±0.005 (temp. of test sample 15° to 35°C; within a range of ±2 pH of the calibration point)
- mV Range: -1200 to 1200.0; -2500 to 2500
- mV Resolution: 0.1; 1.0
- mV Accuracy: ±0.3; ±1.0 (temp. of test sample 15° to 35°C)
- ISE Range (mg/L, mg/kg, mmol/L, ppm, %): 0.000 to 9.999; 10.00 to 99.99; 100.0 to 999.9; 1000 to 999999
- ISE Range (μmol/L): 0.000 to 9.999; 10.00 to 99.99; 100.0 to 999.9; 1000 to 9999
- ISE Resolution (mg/L, mg/kg, mmol/L, ppm, %): 0.001; 0.01; 0.1; 1
- ISE Resolution (μmol/L): 0.001; 0.01; 0.1; 1
- ISE Accuracy: ±0.3; ±1.0 (temp of test sample 15° to 35°C)
- Temperature Range: -5° to +105°C (23° to 221°F)
- Temperature Resolution: 0.1
- Temperature Accuracy: ±0.1
- Memory: 5000 data sets auto logging, 500 data sets manual logging
- Manual Temperature Input: -25° to +130°C (-13° to +266°F) in increments of 1
- Storage Temperature: -25° to +65°C (-13° to +149°F)
- Unit of Measure: Sample subtraction, Known addition (double), Sample addition, ISE Measurements: Blank value correction, increment methods: Known addition (single), Known subtraction
- Weight: 1kg (2 lb.)
- Dimensions (L x W x H): 28 x 23 x 8cm (11 x 9 x 3.2 in.)
Order Info
Cables, probes/sensors, and accessories sold separately.
Certifications
IP43, electrical protective class III, cETLus, CE
Compliance
GLP/AQA compliant
Warranty and Services
Three years
Specifications
±0.3; ±1.0 (temp. of test sample 15° to 35°C) | |
±0.1 | |
4 x alkaline AA batteries (also compatible with four NiMH AA rechargeables) | |
CE, cETLus; electrical protective class III; IP43 | |
pH/ISE 1320P Laboratory ISE Benchtop Meter with integrated Printer | |
0.000 to 9.999, 10.00 to 99.99, 100.0 to 999.9, 1000 to 999999 μmol/L, 0.000 to 9.999, 10.00 to 99.99, 100.0 to 999.9, 1000 to 9999 in mg/L, mg/kg, mmol/L, ppm, % | |
-2.0 to 20.0, -2.00 to 20.00, -2.000 to 19.999 | |
0.1, 0.01, 0.001 | |
Color Graphic Display | |
Meter with integrated printer, universal power supply, electrode stand, manual, PC software and USB cable | |
Four alkaline AA batteries fully charged provides approx.150 hours of illumination; universal power supply; also compatible with four NiMH AA rechargeables | |
0.1 | |
3 Years |
±0.1; ±0.01; ±0.005 (temp. of test sample 15° to 35°C; within a range o | |
approx.150 hours with illumination, 1000 hours without | |
1, 2, 3, 4 or 5 point with or without autobuffer recognition; USA and NIST and 22 pre-programmed buf | |
5000 data sets auto logging, 500 data sets manual logging | |
2 to 7 | |
-1200 to 1200.0, -2500 to 2500 | |
0.001, 0.01, 0.1, 1 μmol/L, 0.001, 0.01, 0.1, 1 in mg/L, mg/kg, mmol/L, ppm, % | |
-5°C to +105°C | |
Measure pH, mV (ORP/Redox); Temperature; ISEs (Ammonia, Ammonium, Bromide, Cadmium, Calcium, Chloride, Copper, Cyanide, Fluoride, Iodide, Lead, Nitrate, Potassium, Silver/Sulfide, Sodium) | |
Dual channel | |
0.1; 1.0 | |
pH/ISE 1320P Laboratory ISE Benchtop Meter with integrated Printer | |
Universal power supply |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction