Learn More
Biocare Medical, LLC XM Factor - 6.0 ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Biocare Medical, LLC XMF963G
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The XM Factor is a potent blocker for eliminating endogenous mouse IgG in mouse tissues. It has been specially designed to work with Biocare Rabbit-on-Rodent and Goat Polymer detection systems. XM Factor cannot be used with Mouse-on-Mouse AP or HRP polymer. By adding XM Factor to the polymer detection, endogenous mouse IgG is dramatically reduced and/or is completely eliminated. Biocare has also developed Rodent Block M that blocks for nonspecific background staining, and at the same time, also helps reduce endogenous mouse IgG. The combination of Rodent Block M and the XM Factor provide clean staining for mouse tissue protocols. It can also be used on an automated staining system.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.