Learn More
TARGETMOL CHEMICALS INC Wortmannin 5mg
Supplier: TARGETMOL CHEMICALS INC T62835MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Wortmannin is the first described PI3K inhibitor with IC50 of 3 nM, with little selectivity within the PI3K family. Also blocks autophagosome formation and potently inhibits DNA-PK/ATM with IC50 of 16 nM and 150 nM.