Learn More
Bulwark™ CP Chemical Splash Protection Lab Coats
Description
- Featuring Milliken ShieldCSR Chemical Splash protection finish resists penetration and repels inadvertent liquid chemical splash
- Snap front closure for easy on/off
- Lay-down collar
- Knit rib cuffs for better fit at wrist
- Splash resistant side pass-throughs for easy inner pocket access
- Pockets include one chest pocket on left chest with a pencil slot and two large lower front pockets with flaps
Specifications
Product Type | Chemical Resistant Lab Coat |
Gender | Unisex |
Color | White |
Disposable | Reusable |
Sterility | Non-sterile |
Resistance | Chemicals |