Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Whatman™ Anotop™ 10 Sterile Syringe Filters
Hold-up volume: <30μL (10mm size), <200μL (25mm)
Supplier: Cytiva 68091102
Description
Hold-up volume: <30μL (10mm size), <200μL (25mm)
Similar to Anotop filters, but include binder-free, glass microfiber prefilter with Anotop™ inorganic membrane in a polypropylene housing.
Glass Microfiber Prefilter
- Increases throughput
- Maintains membrane performance
- Reduces need for centrifuging before filtering, sequential filtering, or refiltering
- Permits use of 0.1μm pore size Anotop Plus filters to remove mycoplasma contaminants from tissue culture
- Hold-up volume: <30μL (10mm size), <200μL (25mm)
- Pressure limit, 7kg/cm (100psi)
Specifications
Anotop 10 mm Sterile Syringe Filters, 0.02 μm (50 pcs) | |
Anapore | |
0.02 μm | |
0.78 cm2 | |
100 mL | |
Female luer lock | |
6.9 bar (100 psi) | |
113°F | |
Polypropylene | |
<30 μL (Hold-Up Volume 10 mm Size), <200 μL (Hold-Up Volume 25 mm Size) |
Single-use | |
Inorganic | |
Sterile | |
HPLC aqueous/organic sample preparation; cold sterilization of growth media; solvent filtration for spectroanalysis; phage and virus filtration; liposome extrusion; removal of high molecular weight proteins or polymers; colloid removal from deionized water | |
Polypropylene (PP) | |
Male luer | |
50 Pieces | |
45°C | |
Syringe Filter | |
10 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction