Learn More
Fisherbrand™ Wet-Strengthened Qualitative Filter Paper Circles
Coarse cellulose fiber papers remove particles >25μm
 View more versions of this product
Catalog No. 09-790-12D
Add to Cart
Description
Description
Coarse cellulose fiber papers remove particles >25μm
  • Creped surface
  • Fast flowrate
  • Suitable for vacuum filtration
Not for nitrogen analysis.
Specifications
Specifications
Disc
>25 μm
Use for coarse particles and gelatinous precipitates.
Coarse
Cellulose
Fast
P8
11 cm
Product Suggestions
Product Suggestions