Learn More
Terumo BCT Inc SCD IIB Weld Integrity Test Kit
Supplier:  Terumo BCT Inc 62002
Encompass
Simplify the process of satisfying quality assurance and Good Manufacturing Practices (GMP) requirements. Perform test welds on-site without shipping your welder. Return your test welds to us to evaluate alignment, leakage, and tensile strength
Catalog No. NC1711320
Explore available promotions