Learn More
Key Scientific Products WEE-TAB ONPG, 28/PK
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Key Scientific Products K1490
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Differentiate slow lactose fermenters and non-fermenters. Demonstration of beta-galactosidase accomplished by hydrolization of substrate that liberates ortho-nitrophenol for a characteristic yellow color. Pack of 28 tablets per tube.