Learn More
Growcells WATER DEPC TREATED 500ML CS20
Supplier: Growcells UPW050020
Molecular Biology Grade/Nuclease Free Water; 0.001% DEPC treated, 18.2 megOhm, 0.1um sterile filtered and autoclaved in TriForest polycarbonate bottles.