Learn More
Invitrogen™ VorTemp™ 56 Shaking Incubator and Temperature Validation Kit
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Description
The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.
VorTemp features:
• A microprocessor to control all parameters including temperature and timed operation
• Self-diagnostic software to continuously monitor for proper temperature and shaking speed, alerting the user to any errors
• Heating and shaking that are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations
• Sturdy construction for stability during high speed shaking
• An insulated chamber to maintain temperature and reduce operating noise
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Content And Storage | • 1 VorTemp 56 Shaking Incubator • 1 QuantiGene Temperature Validation Kit |
Material | Coated Steel |
Relative Humidity | 85% |
Temperature Range (Metric) | 4°C to 99.5°C |
Type | Shaking Incubator |
Product Line | VorTemp™ |
Shipping Condition | Room Temperature |
Voltage | 220 V |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.