Learn More
Sklar Instruments Curette, Volkmann Bone Curette, Sklar 6-3/4 in., #000, Oval, Straight, Sharp/Blunt, Stainless Steel, EA.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Sklar Instruments 407141
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Curette, Volkmann Bone Curette, Sklar, 6-3/4 in., #000, Oval, Straight, Sharp/Blunt, Stainless Steel, EA. Sklar's Volkman Bone Curette is a debulking instrument commonly used in orthopedic procedures. It is used to scrape away and remove fibrous tissue and other debris from bone. It may also be used to scrape and remove small portions of the bone itself. Volkmann bone curettes are available as single or double-ended instruments, in various lengths, with multiple tip configurations. This is the #000 curette and has one oval scoop tip measuring 3.6mm diameter. The working edge is sharp. This product is straight with a flat, smooth handle that is approximately 10mm wide and gradually tapers to the tips. It has an overall length of 6-3/4 inches. Lifetime warranty against all manufacturer defects.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.