Learn More
Perkin Elmer US LLC Volatile Aluminum Sample Pans and Covers - Qty. 400
Supplier: Perkin Elmer US LLC 02190062
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For use with Autosamplers, these pans are used to prepare sample materials for your DSC Autosampler. These pans and covers are hermetically sealed and may also be used for non-robotic systems.