Learn More
Perkin Elmer US LLC Voaltile Sample Capsule - Pan
Supplier: Perkin Elmer US LLC 03191525
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
These high pressure pans and covers contain a sample in a sealed environment throughout an experiment. This ensures that the heat generated from vaporization or a volatile reaction does not interfere with the analyzer's measurements.