Learn More
Glass Expansion Viton Pump Tube 2tag 1.02mm ID White/White (PKT 12)
Supplier:  Glass Expansion VS1.02WHT
Encompass_Preferred
Viton Pump Tube 2tag 1.02mm ID White/White (PKT 12).Tag Colors: White/WhiteID mm: 1.02Wall Thickness mm: 0.85OD mm 2.72Overall Length: 175mm (127mm spacing between tags)Number of Tubes Per Pack: 12Physical Properties: Fluoropolymer elastomerSpecial Properties: Special tubing for concentrated acids and corrosive solventsMaterial: Made from Fluran HC F-5500-A or equivalent.Limitations: Limited service lifeGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2922