Learn More
Glass Expansion Viton Pump Tube 2tag 0.76mm ID Black/Black (PKT 12)
Supplier:  Glass Expansion VS0.76BLK
Encompass_Preferred
Viton Pump Tube 2tag 0.76mm ID Black/Black (PKT 12).Tag Colors: Black/BlackID mm: 0.76Wall Thickness mm: 0.85OD mm 2.46Overall Length: 175mm (127mm spacing between tags)Number of Tubes Per Pack: 12Physical Properties: Fluoropolymer elastomerSpecial Properties: Special tubing for concentrated acids and corrosive solventsMaterial: Made from Fluran HC F-5500-A or equivalent.Limitations: Limited service lifeGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2923