Learn More
Kimberly-Clark™ Professional Visors for BC100 Bump Caps
Kimberly-Clark™ Professional Visors for BC100 Bump Caps provide on-the-job protection for tasks that produce dust and splashes, as in light industrial manufacturing, landscaping and food processing.
Supplier: Kimberly-Clark™ Professional 28765
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Description
- Compatible with the Hard Hat Interchange System
- Snaps into place on compatible head gear and hard hats without removing hard hat – no special tools required
- When used with any Jackson Safety Brand Headgear, the faceshield meets the performance requirements of ANSI Z87.1
Specifications
PETG (thermoplastic polymer resin) |