Learn More
TWD Scientific LLC BioX 750uL SBS Format Minitube Combo. Case of 1920 Tubes and 20 Racks.
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: TWD Scientific LLC BX894283
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
BioX 750µL SBS format screw top minitubes with blue caps. Vials come loaded into a barcoded 96-well rack with square wells to eliminate slip. The internally threaded cryovials feature 2D barcodes on the bottom and are gamma radiation sterilized. 96 tubes/rack, 20 racks/case. price/case.
This item is not returnable.
View return policy