Learn More
World Precision Instrument Butyl Cyanoacrylate, Low Toxicity Adhesive
Supplier: World Precision Instrument VETBOND
Biocompatible adhesive that is four times stronger than butyl cyanoacrylate and less toxic
Cyanoacrylate adhesives have been on the market since 1958. Most industrial or household grade cyanoacrylate is made of shorter alkyl chain derivatives such as methyl or ethyl cyanoacrylate (WPI #7341 and #7342).