Learn More
Air Science Usa Vertical Clean Bench 24" Wide with UV System
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Air Science Usa VLF24A(UV)
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Vertical Laminar Flow Cabinet 24" Wide With UV Lamp and front cover. Protects your non-hazardous applications with Vertical unidirectional (laminar) ISO Class 4 air across the work area. Includes Stainless Steel Workurface. CART-BW-VLF-24 Base Stand avilable and sold separatly.