Learn More
MCR Safety Verdict Goggle
Click to view available options
Description:
Foam-lined
Not foam lined
Description
- Protective PVC body permits a comfortable fit over prescription eyewear
- Optional soft foam lining cushions the face and improves the seal
- Offered in both scratch-resistant polycarbonate lens and anti-fog lens
- Available with either a wide elastic headband or a neoprene headband
- Meets or exceeds ANSI Z87+ and CSA Z94.3.
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Type | Duramass™ Hard Coat |
Description | Foam-lined |
Lens Tint | Clear |
Frame Color | Smoke |
Ventilation Details | Indirect vent |