Learn More
Foxx Life Sciences EZwaste 110 HPLC Solvent Waste System, ¼-28 Thread Size Replacement Chemical Exhaust Filter with Indicator, Activated Carbon, 5/CS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Foxx Life Sciences 2502304FLS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Universal ¼ -28 connection exhaust vent filter rids the ambient air of volatile compounds known to adversely affect health and safety. The large capacity 65g absorbent component, filters out nearly all evaporating organic solvent vapors. The optional qualitative colorimetric indicator recognizes trace amounts of acetone, acetonitrile, aliphatic and aromatic hydrocarbons, ethanol, ethyl acetate, methanol, methylene chloride, and THF. Designed to provide real-time indication of the breakthrough of vapors, when the absorbent is saturated, a color change in the detector from orange/pink to dark purple. A small charcoal filter absorbs the vapors after they pass the detector making this both an ideal and necessary saftey additon to the laboratory.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.