Learn More
Hampton Research VDX PLATE W/SEALANT,40/ CASE
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Hampton Research HR3171
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
24 well crystallization plate for hanging drop or sitting drop vapor diffusion crystallization (when used with Micro-Bridges) or dialysis crystallization (when used with Dialysis Buttons). Stackable, optically clear plastic 24 well plates with raised covers (to allow room for cover slides) and flat bottoms for exceptional optics. Raised, wide rings around each reservoir (well) minimize cross contamination and allow each well to be individually sealed with 22 mm diameter circle or square cover slides. Plates are individually wrapped and supplied with applied sealant.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.