Learn More
Cole-Parmer™ VapLock™ Inlet Check Valves for Solvent Management
VapLock™ Inlet Check Valves for Solvent Management maintain pressure equilibrium within the bottle for safe solvent handling
Supplier: Cole-Parmer™ 1202053
Description
- Minimize evaporation and relieve pressure to improve safety
- Allow air into supply container to displace vacuum and keep solvent flowing
Warranty and Services
None
Specifications
PTFE, EPDM | |
VapLock Solvent Containment Systems | |
1/4 in.-28 UNF(M) |
Solvent Safety | |
Inlet Check Valve; 0.1-0.25 psi crack pressure | |
1 Ea. |