Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Semi-Rigid Tubing, UNF(M)
VapLock™ Adapters for Semi-Rigid Tubing, UNF(M) are designed for secure liquid-tight connections
Supplier: Cole-Parmer™ 1202007
Description
- Color-coded for easy identification
- Features two-piece nut and ferrule system
Warranty and Services
None
Specifications
1/4 in.-28 UNF(M) | |
VapLock Solvent Containment Systems | |
3 mm OD Compression x UNF(M) |
Solvent Safety | |
Yellow / Orange PP / ETFE | |
Tubing Adapter, 3.0 mm OD x 1/4-28 UNF(M), yellow PP w/ orange ETFE; 10/Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction