Learn More
Cole-Parmer™ VapLock™ Adapters for Flexible Tubing
VapLock™ Adapters for Flexible Tubing are designed for secure leak-resistant connections
Supplier: Cole-Parmer™ 1202033
Description
Use with flexible, soft-wall tubingWarranty and Services
None
Specifications
Solvent Safety | |
Gray Polypropylene | |
1/2 in. Barb x NPT(M) |
VapLock Solvent Containment Systems | |
Tubing Adapter, 1/2 in. ID x 1/2 in. NPT(M), Polypropylene, 10/Pk. | |
1/2 in. NPT(M) |