Learn More
Chemglass Life Sciences Valve, Hi-Vac, 1-Arm, 0-20mm Bore, Viton O-Ring
Supplier: Chemglass Life Sciences CG53404
Valve, Hi-Vac, 1-Arm, 0-20mm Bore, Viton O-Ring. CHEM-VAC valve has single sidearm sealed at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Valve has a TFE plug with three o-rings, the tip o-ring making the primary seal on the precision formed tapered seat. O-rings on the non-rotating shaft form an excellent dynamic seal between the TFE plug and glass barrel. See CG-935 for replacement plug, CG-537 for o-ring replacement kit, and CG-902 for glass portion only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.