Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Value Convertible Refrigerator/Freezer PROMO
Flexible and economical laboratory storage.
Description
Fisherbrand™ Value Convertible Refrigerator/Freezers offer economical and simple sample storage while also offering increased flexibility of operating as either a refrigerator or a freezer with the push of a button.
Note: Comes configured as a refrigerator
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
General Purpose Refrigerator (Convertible to Freezer) | |
594 L | |
1°C to 12°C | |
Hinged, Open Left to Right, Non-reversible | |
3 Adjustable | |
Bottom-mounted | |
2 - Refrigerator or Freezer | |
White-enameled Steel | |
ABS Plastic | |
White | |
Indoor general storage of non-critical samples and reagents | |
4 in. on sides, 4 in. at back, 11 in. on top | |
13 months parts and labor | |
57.15 x 67.3 x 175.26 cm (22.5 x 26.5 x 69 in.) | |
NEMA 5-15 P | |
115 V/60 Hz | |
244 lb. |
21 cu. ft. | |
-12°C to -24°C | |
Adjustable | |
1 (1 compartment) | |
4 fixed shelves | |
Automatic | |
R600a | |
Cyclopentane | |
2 adjustable leveling feet | |
Electronic control | |
Interior | |
10 years | |
73.5 x 83.2 x 195 cm (28.94 x 32.76 x 76.77 in.) | |
2.2 A | |
115 V | |
50 Hz | |
110.6 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction